Novus Biologicals™ Lightning-Link™ Rapid Atto 700 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3540015

 View more versions of this product

Catalog No. 35-400-15

  Check Availability
Add to Cart

    Specifications

    Product Type Rapid Atto 700 Antibody Labeling Kit
    Conjugate Atto700
    Quantity 1 x 2mg Reaction
    Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Atto700 is one of a new generation of fluorescent labels that emits in the red spectral region. It has a strong absorption peak at 690nm, with an extinction coefficient of 1.2 x 105 cm-1M-1, and a maximum emission peak at 725nm.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit