Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Novus Biologicals™ Lightning-Link™ Rapid Atto 700 Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3540030
 View more versions of this product
Catalog No. 35-400-30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Atto700 is one of a new generation of fluorescent labels that emits in the red spectral region. It has a strong absorption peak at 690nm, with an extinction coefficient of 1.2 x 105 cm-1M-1, and a maximum emission peak at 725nm.
Order Info
Quantity
Description & Specifications
Specifications
3 x 20μg Reactions
Store at -20°C. Avoid freeze-thaw cycles.
Atto700
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Rapid Atto 700 Antibody Labeling Kit