Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto 700 Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3540010
 View more versions of this product
Catalog No. 35-400-10
  Check Availability
Add to cart
Specifications
Description
Atto700 is one of a new generation of fluorescent labels that emits in the red spectral region. It has a strong absorption peak at 690nm, with an extinction coefficient of 1.2 x 105 cm-1M-1, and a maximum emission peak at 725nm.
Order Info
Quantity
Description & Specifications
Specifications
3 x 200μg Reactions
Store at -20°C. Avoid freeze-thaw cycles.
Atto700
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Rapid Atto 700 Antibody Labeling Kit