Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto390 Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3490015
 View more versions of this product
Catalog No. 34-900-15
  Check Availability
Add to cart
Specifications
Description
Atto390 is one of a new generation of fluorescent labels with a coumarin structure. It has a strong absorption at 390nm, high fluorescence at 479nm (extinction coefficient 2.4 x10 cm-1M-1) and high quantum yield.
Order Info
Quantity
Description & Specifications
Specifications
1 x 2mg Reaction
Store at -20°C. Avoid freeze-thaw cycles.
Rapid Atto390 Antibody Labeling Kit