Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto390 Antibody Labeling Kit
Antibody labeling kit
$378.35 - $550.85
Specifications
Product Type Rapid Atto390 Antibody Labeling Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
34-900-30
View Documents
Novus Biologicals™
3490030
3 x 20μg Reactions
Each for $378.35
Add to cart
34-900-10
View Documents
Novus Biologicals™
3490010
3 x 200μg Reactions
Each for $550.85
Add to cart
Description
Description
Atto390 is one of a new generation of fluorescent labels with a coumarin structure. It has a strong absorption at 390nm, high fluorescence at 479nm (extinction coefficient 2.4 x10 cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid Atto390 Antibody Labeling Kit