Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto488 Antibody Labeling Kit
Applications: Flow Cytometry
$378.35 - $953.35
Specifications
Product Type Rapid Atto488 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
35-000-30
View Documents
Novus Biologicals™
3500030
3 x 20μg Reactions Each for $378.35
Add to cart
35-000-10
View Documents
Novus Biologicals™
3500010
3 x 200μg Reactions Each for $550.85
Add to cart
35-000-15
View Documents
Novus Biologicals™
3500015
1 x 2mg Reaction Each for $953.35
Add to cart
Description
Description
Atto488 is one of a new generation of fluorescent labels, which has been optimised for excitation with an argon laser. It has a strong absorption at 501nm, high fluorescence at 523nm (extinction coefficient 9.0 x10 cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid Atto488 Antibody Labeling Kit