Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto565 Antibody Labeling Kit
Applications: Flow Cytometry
$378.35 - $550.85
Specifications
Product Type Rapid Atto565 Antibody Labeling Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
35-100-30
View Documents
Novus Biologicals™
3510030
3 x 20μg Reactions
Each for $378.35
Add to cart
35-100-10
View Documents
Novus Biologicals™
3510010
3 x 200μg Reactions
Each for $550.85
Add to cart
Description
Description
Atto565 is one of a new generation of fluorescent labels. It has a strong absorption at 563nm, high fluorescence at 592nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield.
Specifications
Specifications
Rapid Atto565 Antibody Labeling Kit