Novus Biologicals™ Lightning-Link™ Rapid Atto565 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3510015

 View more versions of this product

Catalog No. 35-100-15

  Check Availability
Add to Cart

    Specifications

    Product Type Rapid Atto565 Antibody Labeling Kit
    Conjugate Atto565
    Quantity 1 x 2mg Reaction
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Atto565 is one of a new generation of fluorescent labels. It has a strong absorption at 563nm, high fluorescence at 592nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit