Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto594 Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3520030
 View more versions of this product
Catalog No. 35-200-30
  Check Availability
Add to cart
Specifications
Description
Atto594 is a fluorescent label belonging to the Rhodamine class of dyes. The dye has a strong absorption at 601nm, high fluorescence at 627nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield (0.85).
Order Info
Quantity
Description & Specifications
Specifications
3 x 20μg Reactions
Store at -20°C. Avoid freeze-thaw cycles.
Atto594
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Rapid Atto594 Antibody Labeling Kit