Novus Biologicals™ Lightning-Link™ Rapid Atto594 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3520010

 View more versions of this product

Catalog No. 35-200-10

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Rapid Atto594 Antibody Labeling Kit
    Conjugate Atto594
    Quantity 3 x 200μg Reactions
    Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Atto594 is a fluorescent label belonging to the Rhodamine class of dyes. The dye has a strong absorption at 601nm, high fluorescence at 627nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield (0.85).

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit