Learn More
Novus Biologicals™ Lightning-Link™ Rapid Atto633 Antibody Labeling Kit
Antibody labeling kit
$529.50 - $1172.00
Specifications
Product Type Rapid Atto633 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
35-300-30
View Documents
Novus Biologicals™
3530030
30 μg
Each for $529.50
Add to cart
35-300-10
View Documents
Novus Biologicals™
3530010
300 μg
Each for $724.00
Add to cart
35-300-15
View Documents
Novus Biologicals™
3530015
1 mg
Each for $1,172.00
Add to cart
Description
Description
Atto633 is one of a new generation of fluorescent labels. It has a strong absorption at 629nm, high fluorescence at 657nm (extinction coefficient 1.3 x105 cm-1M-1) and high quantum yield. Atto633 can be used as a substitute dye for DY-633, LightCycler Red 640, Alexa Fluor 633, and Cy 5.
Specifications
Specifications
Rapid Atto633 Antibody Labeling Kit