Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Cy3 Antibody Labeling Kit
Antibody labeling kit
Supplier: Novus Biologicals™ 3400030
Description
Cyanine Dye 3 (Cy3) has a strong absorption peak at 550nm and a secondary absorption peak at 520nm. The maximum emission peak is at 570nm, with an excitation coefficient of 1.5 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;. Cy3 can be used as an alternative to Alexa Fluor 555, DY-550, Rhodamine Red-X and TAMRA [5(6)-Carboxytetramethylrhodamine].Order Info
Quantity
Specifications
Cy3 | |
30 μg | |
Rapid Cy3 Antibody Labeling Kit |
Store at -20°C. Avoid freeze-thaw cycles. | |
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |