Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Cy3 Antibody Labeling Kit
Antibody labeling kit
$542.50 - $1147.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | Cy3 |
Kit Contents | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
Product Type | Rapid Cy3 Antibody Labeling Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
34-000-30
|
Novus Biologicals™
3400030 |
30 μg |
Each for $542.50
|
|
|||||
34-000-10
|
Novus Biologicals™
3400010 |
300 μg |
Each for $760.00
|
|
|||||
34-000-15
|
Novus Biologicals™
3400015 |
1 mg |
Each for $1,147.00
|
|
|||||
Description
Cyanine Dye 3 (Cy3) has a strong absorption peak at 550nm and a secondary absorption peak at 520nm. The maximum emission peak is at 570nm, with an excitation coefficient of 1.5 x105cm-1M-1. Cy3 can be used as an alternative to Alexa Fluor 555, DY-550, Rhodamine Red-X and TAMRA [5(6)-Carboxytetramethylrhodamine].Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Cy3 | |
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent | |
Rapid Cy3 Antibody Labeling Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title