Novus Biologicals™ Lightning-Link™ Rapid Cy5.5 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3430015

 View more versions of this product

Catalog No. 34-300-15

  Check Availability
Add to Cart

    Specifications

    Product Type Rapid Cy5.5 Antibody Labeling Kit
    Conjugate Cy5.5
    Quantity 1 x 2mg Reaction
    Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Cyanine Dye 5.5 (Cy5.5) has a strong absorption peak at 675nm. The maximum emission peak is at 694nm, with an extinction coefficient of 2.5 x105cm-1M-1. Cy5.5 can be used as an alternative to Alexa Fluor 680, WellRed DP-3A, DY- 676 and LightCycler Red.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit