Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 350 Antibody Labeling Kit
Antibody labeling kit
$378.35 - $918.85
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
32-000-30
View Documents
Novus Biologicals™
3200030
N/A 3 x 20µg Reactions
Each for $378.35
Add to cart
32-000-15
View Documents
Novus Biologicals™
3200015
N/A 1 x 2mg Reaction
Each for $918.85
Add to cart
32-000-10
View Documents
Novus Biologicals™
3200010
N/A 3 x 200µg Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
DyLight 350 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 353nm, high fluorescence at 432nm (extinction coefficient 1.5 x104cm-1M-1) and high quantum yield.