Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 350 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3200010

 View more versions of this product

Catalog No. 32-000-10

  Check Availability
Add to Cart

    Specifications

    Product Type Rapid DyLight 350 Antibody Labeling Kit
    Conjugate DyLight 350
    Quantity 3 x 200μg Reactions
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    DyLight 350 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 353nm, high fluorescence at 432nm (extinction coefficient 1.5 x104cm-1M-1) and high quantum yield.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit