Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 405 Antibody Labeling Kit
Antibody labeling kit
$550.85 - $895.85
Specifications
Product Type Rapid DyLight 405 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-100-10
View Documents
Novus Biologicals™
3210010
3 x 200μg Reactions Each for $550.85
Add to cart
32-100-15
View Documents
Novus Biologicals™
3210015
1 x 2mg Reaction Each for $895.85
Add to cart
32-100-30
View Documents
Novus Biologicals™
3210030
3 x 20μg Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
DyLight 405 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 400nm, high fluorescence at 420nm (extinction coefficient 3.0 x104 cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 405 Antibody Labeling Kit