Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 488 Antibody Labeling Kit
Applications: Flow Cytometry
$378.35 - $895.85
Specifications
Product Type Rapid DyLight 488 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-200-30
View Documents
Novus Biologicals™
3220030
3 x 20μg Reactions
Each for $378.35
Add to cart
32-200-10
View Documents
Novus Biologicals™
3220010
3 x 200μg Reactions
Each for $550.85
Add to cart
32-200-15
View Documents
Novus Biologicals™
3220015
1 x 2mg Reaction
Each for $895.85
Add to cart
Description
Description
DyLight 488 provides green fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 493nm, high fluorescence at 518nm (extinction coefficient 7.0 x104cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 488 Antibody Labeling Kit