Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 594 Antibody Labeling Kit
Antibody labeling kit
$378.35 - $895.85
Specifications
Product Type Rapid DyLight 594 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-400-30
View Documents
Novus Biologicals™
3240030
3 x 20μg Reactions Each for $378.35
Add to cart
32-400-10
View Documents
Novus Biologicals™
3240010
3 x 200μg Reactions Each for $550.85
Add to cart
32-400-15
View Documents
Novus Biologicals™
3240015
1 x 2mg Reaction Each for $895.85
Add to cart
Description
Description
DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x104 cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 594 Antibody Labeling Kit