Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 633 Antibody Labeling Kit

Antibody labeling kit

$378.35 - $895.85

Specifications

Product Type Rapid DyLight 633 Antibody Labeling Kit
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Item Description DyLight™ 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10cm-1M-1) and high quantum yield.
Conjugate DyLight 633
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-500-30
View Documents
Novus Biologicals™
3250030
3 x 20μg Reactions Each for $378.35
Add to cart
 
32-500-10
View Documents
Novus Biologicals™
3250010
3 x 200μg Reactions Each for $550.85
Add to cart
 
32-500-15
View Documents
Novus Biologicals™
3250015
1 x 2mg Reaction Each for $895.85
Add to cart
 
Description

Description

DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10 cm-1M-1) and high quantum yield.
  • Lightning-Link™ Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Easy to use antibody labeling kit with quick labeling times
  • From 10µg up to 2mg
  • Kit with 30 seconds hands-on time
  • No separation steps and no losses
  • Scalable technology
Applications:
  • Western Blotting, ELISA, Immunohistochemistry, Immunofluorescence and FACS analysis
Specifications

Specifications

Rapid DyLight 633 Antibody Labeling Kit
Store at -20°C. Avoid freeze-thaw cycles.
DyLight™ 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10cm-1M-1) and high quantum yield.
DyLight 633
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit