Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 650 Antibody Labeling Kit
Applications: Flow Cytometry
$378.35 - $895.85
Specifications
Product Type Rapid DyLight 650 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-600-30
View Documents
Novus Biologicals™
3260030
3 x 20μg Reactions
Each for $378.35
Add to cart
32-600-10
View Documents
Novus Biologicals™
3260010
3 x 200μg Reactions
Each for $550.85
Add to cart
32-600-15
View Documents
Novus Biologicals™
3260015
1 x 2mg Reaction
Each for $895.85
Add to cart
Description
Description
DyLight 650 generates far red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 652nm, high fluorescence at 672nm (extinction coefficient 2.5 x105cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 650 Antibody Labeling Kit