Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 680 Antibody Labeling Kit
Antibody labeling kit
$378.35 - $895.85
Specifications
Product Type Rapid DyLight 680 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-700-30
View Documents
Novus Biologicals™
3270030
3 x 20μg Reactions Each for $378.35
Add to cart
32-700-10
View Documents
Novus Biologicals™
3270010
3 x 200μg Reactions Each for $550.85
Add to cart
32-700-15
View Documents
Novus Biologicals™
3270015
1 x 2mg Reaction Each for $895.85
Add to cart
Description
Description
DyLight 680 provides red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 682nm, high fluorescence at 715nm (extinction coefficient 1.4 x105cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 680 Antibody Labeling Kit