Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 755 Antibody Labeling Kit
Antibody labeling kit
$511.50 - $1094.00
Specifications
Product Type Rapid DyLight 755 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-800-30
View Documents
Novus Biologicals™
3280030
30 μg
Each for $511.50
Add to cart
32-800-10
View Documents
Novus Biologicals™
3280010
300 μg
Each for $705.00
Add to cart
32-800-15
View Documents
Novus Biologicals™
3280015
1 mg
Each for $1,094.00
Add to cart
Description
Description
DyLight 755 provides far red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 754nm, high fluorescence at 776nm (extinction coefficient 2.2 x105cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 755 Antibody Labeling Kit