Learn More
Novus Biologicals™ Lightning-Link™ Rapid DyLight™ 800 Antibody Labeling Kit
Antibody labeling kit
$378.35 - $895.85
Specifications
Product Type Rapid DyLight 800 Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
32-900-30
View Documents
Novus Biologicals™
3290030
3 x 20μg Reactions
Each for $378.35
Add to cart
32-900-10
View Documents
Novus Biologicals™
3290010
3 x 200μg Reactions
Each for $550.85
Add to cart
32-900-15
View Documents
Novus Biologicals™
3290015
1 x 2mg Reaction
Each for $895.85
Add to cart
Description
Description
DyLight 800 provides near infrared fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 770nm, high fluorescence at 794nm (extinction coefficient 2.7 x105cm-1M-1) and high quantum yield.
Specifications
Specifications
Rapid DyLight 800 Antibody Labeling Kit