Novus Biologicals™ Lightning-Link™ Rapid FluoProbe™594 Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3610015

 View more versions of this product

Catalog No. 36-100-15

  Check Availability
Add to Cart

    Specifications

    Product Type Rapid FluoProbe594 Antibody Labeling Kit
    Conjugate FluoProbes 594
    Quantity 1 x 2mg Reaction
    Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    FluoProbes 594 is one of a new generation of fluorescent labels designed for labelling biomolecules. It has a strong absorption at 591nm, high fluorescence at 616nm (extinction coefficient 9.2 x 104 cm-1M-1) and high quantum yield. FluoProbes 594 can be used as a substitute dye for BODIPY TR-x, Carboxynaphofluorescein and Alexa Fluor 594.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit