Learn More
Novus Biologicals™ Lightning-Link™ Rapid FluoProbe™594 Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3610010
 View more versions of this product
Catalog No. 36-100-10
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
FluoProbes 594 is one of a new generation of fluorescent labels designed for labelling biomolecules. It has a strong absorption at 591nm, high fluorescence at 616nm (extinction coefficient 9.2 x 104 cm-1M-1) and high quantum yield. FluoProbes 594 can be used as a substitute dye for BODIPY TR-x, Carboxynaphofluorescein and Alexa Fluor 594.
Order Info
Quantity
Description & Specifications
Specifications
3 x 200μg Reactions
Store at -20°C. Avoid freeze-thaw cycles.
FluoProbes 594
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Rapid FluoProbe594 Antibody Labeling Kit
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'