Learn More
Novus Biologicals™ Lightning-Link™ Rapid Fluorescein Antibody Labeling Kit
Antibody labeling kit
$343.85 - $780.85
Specifications
Product Type Rapid Fluorescein Antibody Labeling Kit
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
31-000-30
View Documents
Novus Biologicals™
3100030
3 x 20μg Reactions
Each for $343.85
Add to cart
31-000-10
View Documents
Novus Biologicals™
3100010
3 x 200μg Reactions
Each for $458.85
Add to cart
31-000-15
View Documents
Novus Biologicals™
3100015
1 x 2mg Reaction
Each for $780.85
Add to cart
Description
Description
Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In water it shows an absorption maximum at 494 nm and an emission maximum of 521 nm.
Specifications
Specifications
Rapid Fluorescein Antibody Labeling Kit