Learn More
Novus Biologicals™ Lightning-Link (R) Streptavidin Antibody Labeling Kit
SDP
Antibody labeling kit
Supplier:  Novus Biologicals™ 7080030
 View more versions of this product
Catalog No. 70-800-30
Add to Cart
Description
Description
Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin. The Lightning-Link Streptavidin kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximizing the opportunity for biotin to bind and create a biotin streptavidin complex. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Order Info
Quantity
Specifications
Specifications
Store at -20°C. Avoid freeze-thaw cycles.
30 μg
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Streptavidin Antibody Labeling Kit
Product Suggestions
Product Suggestions