Novus Biologicals™ Lightning-Link™ Type A Biotin Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus Biologicals™ 7040015

 View more versions of this product

Catalog No. 70-400-15

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

For Research Use Only


    Specifications

    Product Type Type A Biotin Antibody Labeling Kit
    Conjugate Biotin
    Quantity 1 x 2mg Reaction
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Lightning-Link Biotin has been optimised for two separate applications. Type A is intended for assays in which a streptavidin-labeled detection reagent will be used, while Type B is optimised for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (ie plates, nitrocellulose, magnetic beads etc).

    Lightning-Link is the easiest and quickest method available for preparing biotinylated antibodies (proteins), requiring just 30 seconds hands-on time, regardless of scale. Biotin with an extended chain is covalently attached in a controlled process at near-neutral pH. Absence of desalting/dialysis steps means that biotinylations can be carried out even at microgram scale without losses. The attachment of biotin to various chemical sites, called biotinylation, is an important laboratory technique, which has been used to study various processes including protein localization, protein interactions, DNA transcription and replication. Biotin binds to the tetrameric avidin proteins, including streptavidin and neutravidin, with very high affinity. This interaction has also been used in various biotechnological applications, such as extraction of biotin tagged proteins and ELISA assays. The amount of antibody used for labeling should be the same as the pack size of LL-Biotin. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile.

    Optimized for assays in which a streptavidin-labeled reagent will be used.

    If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit