Novus Biologicals™ Lightning-Link™ Type B Biotin Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus Biologicals™ 7150030

 View more versions of this product

Catalog No. 71-500-30

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Type B Biotin Antibody Labeling Kit
    Conjugate Biotin
    Quantity 3 x 20μg Reactions
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    The attachment of biotin to biomolecules is an important laboratory technique. Biotin binds to the tetrameric avidin proteins, including streptavidin and neutravidin, with exceptionally high affinity, and this interaction is exploited in various applications such as western blotting, immunohisthochemistry and ELISA. Our biotinylation kits are optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step following the biotinylation reaction, saving you both time and material. Lightning-Link Biotin has been optimised for two separate applications: Type A - intended for assays in which a streptavidin-labeled detection reagent will be used. Type B - optimised for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (ie plates, nitrocellulose, magnetic beads etc). If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit