Learn More
Novus Biologicals™ Lightning-Link™ Type B Biotin Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus Biologicals™ 7150015
 View more versions of this product
Catalog No. 71-500-15
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
The attachment of biotin to biomolecules is an important laboratory technique. Biotin binds to the tetrameric avidin proteins, including streptavidin and neutravidin, with exceptionally high affinity, and this interaction is exploited in various applications such as western blotting, immunohisthochemistry and ELISA. Our biotinylation kits are optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step following the biotinylation reaction, saving you both time and material. Lightning-Link Biotin has been optimised for two separate applications: Type A - intended for assays in which a streptavidin-labeled detection reagent will be used. Type B - optimised for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (ie plates, nitrocellulose, magnetic beads etc).
Order Info
Quantity
Description & Specifications
Specifications
1 x 2mg Reaction
Store at -20°C. Avoid freeze-thaw cycles.
Biotin
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Type B Biotin Antibody Labeling Kit