Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LRPAP Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP178883100UG

Catalog No. NBP17888310


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 35-357 of Human LRPAP The Recombinant Human LRPAP Protein is derived from E. coli. The Recombinant Human LRPAP Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

NP_002328
SDS-Page
4043
SDS-PAGE
100μg
LRPAP1, 35-357aa, Human, His tag, E. coli (NP_002328).
RUO
Unlabeled
1.0mg/mL
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT
40.4kDa
>90%
Recombinant human LRPAP1 protein, fused to His-tag at N-terminus, expressed in E. coli
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
LRPAP
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit