Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lymph Node Tissue Slides (Normal)- Paraffin
SDP

Applications: Immunohistochemistry-Paraffin

Supplier:  Novus Biologicals™ NBP230183

Catalog No. NBP230183


Add to Cart

Description

Description

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications

Specifications

Human
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Normal
Lymph Node Tissue Slides (Normal)
Human
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C. Do not freeze.

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit