Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ M-CSF Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1431510.005MG

 View more versions of this product

Catalog No. NBP14315105


Add to cart

Description

Description

Insect expressed C-terminal His-tagged Lys33-Pro187, Accession #: NM_001113530. Source: Insect The Recombinant Mouse M-CSF Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Flow Cytometry, Immunoassay
0.005mg
Store at -80°C. Avoid freeze-thaw cycles.
M-CSF
Sterile liquid; phosphate buffered saline with 1% BSA, pH 7.2
Mouse
RUO
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit