Learn More
Novus Biologicals™ Mouse Mouse IgG, Fab2 Protein
Mouse Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197018
Catalog No. NBP197018
Add to cart
Description
Description
IgG F(ab)2 Isotype Control antibody specifically detects IgG F(ab)2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page.
Specifications
Specifications
WB, ELISA, IP
Mouse IgG, Fab2 Protein
Mouse
Mouse
Potassium Phosphate, 0.15M Sodium Chloride pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum, anti-Mouse IgG and anti-Mouse IgG F(ab)2. No reaction was observed against anti-Mouse IgG F(c) or anti-Pepsin.