Learn More
Novus Biologicals™ Mouse Mouse IgG, Fc Protein
Mouse Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP196784
Catalog No. NBP196784
Add to cart
Description
Description
IgG Fc Isotype Control antibody specifically detects IgG Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page.
Specifications
Specifications
WB, ELISA, IHC, IHC-Paraffin, IP, SDS-Page
Mouse IgG, Fc Protein
−80°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum, anti-Mouse IgG and anti-Mouse IgG F(c). No reaction was observed against anti-Mouse IgG F(ab)2 or anti-Papain.
0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2
Mouse IgG F(c) fragment
Mouse
Mouse