Learn More
Novus Biologicals™ IgG1 Mouse anti-Mouse, Clone: MG1L, Isotype Control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals™ NBP196982
Catalog No. NBP196982
Add to cart
Description
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, SDS-Page.
Specifications
Specifications
WB, ELISA, FC, IP
Mouse IgG1 Protein
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Mouse igG lambda isotype control is non-reactive with antisera to mouse IgG2a, IgG2b, IgG3 IgM, and IgA. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum.
0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2
Protein A purified
Mouse
Mouse