Learn More
Novus Biologicals™ IgG2a Mouse anti-Mouse, Clone: M2AL, Isotype Control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals™ NBP197031
Catalog No. NBP197031
Add to cart
Description
Description
IgG2a Monoclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
WB, ELISA, IP
Mouse IgG2a Protein
Mouse
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum. By ELISA, the material is non-reactive with antibodies to mouse IgG1, IgG2b, IgG3, IgM and IgA.
0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Protein A purified
Mouse