Learn More
Novus Biologicals™ IgG2a Mouse anti-Mouse, Clone: M2AK, Isotype Control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals™ NBP196981
Catalog No. NBP196981
Add to cart
Description
Description
IgG2a Monoclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, SDS-Page.
Specifications
Specifications
WB, ELISA, FC, IP
Mouse IgG2a Protein
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
In an Ouchterlony double diffusion assay the material is non-reactive with antisera to mouse IgG1, IgG2b, IgG3 IgM and IgA. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum.
0.02 M Potassium Phosphate, 0.5M Sodium Chloride pH 7.2
Affinity Purified
Mouse
Mouse