Learn More
Mouse IgG3 Kappa Light Chain Isotype Control (MG3K), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196978
Description
IgG3 Kappa Light Chain Monoclonal antibody specifically detects IgG3 Kappa Light Chain in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
IgG3 Kappa Light Chain | |
MG3K | |
0.02 M Potassium Phosphate, 0.5M Sodium Chloride pH 7.2 | |
Isotyping assay resulted non-reactive with antisera to mouse IgG1, IgG2a, IgG2b, IgA, IgM |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:1000-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, SDS-Page, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 | |
1 mg | |
4°C short term, −20°C long term. Avoid freeze-thaw cycles. |