Learn More
Novus Biologicals™ IgG3 Mouse anti-Mouse, Clone: MG3L, Isotype Control, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals™ NBP196977
Catalog No. NBP196977
Add to cart
Description
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
WB, ELISA, FC, IP
Mouse IgG3 Protein
IGHG3
Mouse
Mouse
0.02 M Potassium Phosphate, 0.5M Sodium Chloride pH 7.2
Affinity Purified
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
In an Ouchterlony double diffusion assay the material is non-reactive with antisera to mouse IgG1, IgG2a, IgG2b, IgM and IgA. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum.