Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse VEGF Recombinant Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Manufacturer:  Novus Biologicals™ NBP130236

Catalog No. NBP130236


Add to cart

Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 205-324 of Mouse VEGF The Recombinant Mouse VEGF Protein is derived from E. coli. The Recombinant Mouse VEGF Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

NP_001020428
20mM Tris-HCl buffer (pH8.0), 10% glycerol
16.3kDa
Protein
0.05mg
VEGF, 205-324 aa, Mouse, His-tagged, Recombinant, E.coli (NP_001020428).
RUO
Mouse
ELISA, SDS-Page
7422
VEGF Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit