Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MSRB2 Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1508970.1MG

 View more versions of this product

Catalog No. NBP15089701


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-182 of Human MSRB2 The Recombinant Human MSRB2 Protein is derived from E. coli. The Recombinant Human MSRB2 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
Liquid. In 20mM Tris-HCl Buffer (pH 7.5) containing 1mM DTT, 0.1mM PMSF, 10% Glycerol
SDS-PAGE
0.1mg
RUO
Unlabeled
SDS-Page
19.5kDa
>90%
Store at -80°C. Avoid freeze-thaw cycles.
MSRB2
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit