Learn More
Novus Biologicals™ NIH 3T3 Nuclear Histone Prep Cell Lysate
Manufacturer:  Novus Biologicals™ NBP178157
Catalog No. NBP178157
Add to cart
Description
Description
The NIH 3T3 Nuclear Histone Prep Cell Lysate is derived from mouse. It has been validated for the following applications: SDS-Page.
Specifications
Specifications
Bone/Cartilage
Triton Extraction Buffer (1X PBS containing 0.5% Triton X 100 (v/v) and a cocktail of phosphatase and protease inhibitors) with 1X Lithium Dodecyl Sulfate (LDS) Sample Buffer at pH 8.4. BME has not been added.
-80°C. Avoid freeze-thaw cycles.
0.1mg
Protein
Nuclear Histone Prep