Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant GST Epitope Tag His Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Supplier: Novus Biologicals™ NBP130259
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-218 of Schistosoma japonicum GST Epitope Tag The Recombinant Parasite GST Epitope Tag Protein is derived from E. coli. The Recombinant Parasite GST Epitope Tag Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
Bioactivity, SDS-PAGE | |
2944 | |
GST Epitope Tag Protein | |
0.1mg | |
MGSSHHHHHH SSGLVPRGSH MSPILGYWKI KGLVQPTRLL LEYLEEKYEE HLYERDEGDK WRNKKFELGL EFPNLPYYID GDVKLTQSMA IIRYIADKHN MLGGCPKERA EISMLEGAVL DIRYGVSRIA YSKDFETLKV DFLSKLPEML KMFEDRLCHK TYLNGDHVTH PDFMLYDALD VVLYMDPMCL DAFPKLVCFK KRIEAIPQID KYLKSSKYIA WPLQGWQATF GGGDHPPKSD LVPR | |
RUO | |
Non-species Specific | |
>95%, by SDS-PAGE |
PBS (pH 7.4), 10% glycerol | |
28.3kDa | |
Protein | |
E.Coli | |
−80°C. Avoid freeze-thaw cycles. | |
Unconjugated | |
Recombinant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction