Learn More
Novus Biologicals™ Nucleoporin 107 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP176926PEP
Description
A blocking peptide containing 20 amino acids near the center of center of human NUP107. Source: Synthetic (Accession #: P57740) The Nucleoporin 107 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
57122 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-76926. For Further Blocking Peptide Related Protocol, Click Here . | |
NUP107 | |
0.05 mg | |
Synthetic |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
nuclear pore complex protein Nup107, nucleoporin 107 kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
Nucleoporin 107 | |
RUO - research use only | |
20 amino acids near the center of human NUP107 |