Learn More
Novus Biologicals™ NUP160 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176928PEP
Catalog No. NBP176928E
Add to cart
Description
Description
A blocking peptide containing 14 amino acids near the carboxy terminus of human NUP160. Source: Synthetic (Accession #: EAW67881) The NUP160 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
23279
For Blocking the binding of NBP1-76928
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
14 amino acids near the carboxy terminus of human NUP160
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
NUP160