Learn More
Novus Biologicals™ Otubain-1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130186
Catalog No. NBP130186
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-271 of Human Otubain-1 The Recombinant Human Otubain-1 Protein is derived from E. coli. The Recombinant Human Otubain-1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_060140
21mM Tris buffer (pH 8.0), 10% glycerol
33.4kDa
Protein
MGSSHHHHHH SSGLVPRGSH MAAEEPQQQK QEPLGSDSEG VNCLAYDEAI MAQQDRIQQE IAVQNPLVSE RLELSVLYKE YAEDDNIYQQ KIKDLHKKYS YIRKTRPDGN CFYRAFGFSH LEALLDDSKE LQRFKAVSAK SKEDLVSQGF TEFTIEDFHN TFMDLIEQVE KQTSVADLLA SFNDQSTSDY LVVYLRLLTS GYLQRESKFF EHFIEGGRTV KEFCQQEVEP MCKESDHIHI IALAQALSVS IQVEYMDRGE GGTTNPHIFP EGSEPKVYLL YRPGHYDILY K
ELISA, SDS-Page
55611
Otubain-1 Protein
>95%
−80°C. Avoid freeze-thaw cycles.