Learn More
Novus Biologicals™ PARC Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP177222PEP
Description
A blocking peptide containing 15 amino acids near the center of human PARC. Source: Synthetic (Accession #: NP_055904) The PARC Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
23113 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-77222. For Further Blocking Peptide Related Protocol, Click Here . | |
CUL9 | |
0.05 mg | |
Synthetic |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
CUL-9, cullin 9, cullin-9, DKFZp686G1042, H7AP1DKFZp686P2024, p53-associated parkin-like cytoplasmic protein, PARCKIAA0708, parkin-like cytoplasmic p53 binding protein, RP3-330M21.2, UbcH7-associated protein 1 | |
PARC | |
RUO - research use only | |
15 amino acids near the center of human Hrk |