Learn More
Novus Biologicals™ Perilipin-3/TIP47 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176719PEP
Catalog No. NBP176719E
Add to cart
Description
Description
A blocking peptide containing 14 amino acids near the amino terminus of human TIP47. Source: Synthetic (Accession #: O60664) The Perilipin-3/TIP47 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
10226
For Blocking the binding of NBP1-76719
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
14 amino acids near the amino terminus of human TIP47
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
Perilipin-3/TIP47